
Trimethobenzamide hydrochloride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:554-92-7
:
Formula:C₂₁H₂₈N₂O₅·HCl
MDL Number:MFCD00057999
:424.9184 g/mol
EINECS:209-075-6
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:187.0 to 191.0 deg-C
Specification Test Results
Frequently Bought Together