Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Tetrahydro-2H-thiopyran-4-ol
Tetrahydro-2H-thiopyran-4-ol ≥98.0% (by GC)
Tetrahydro-2H-thiopyran-4-ol ≥98.0% (by GC)
:
TCI
Tetrahydro-2H-thiopyran-4-ol ≥98.0% (by GC)
:
TCI
:
29683-23-6
:
:
C₅H₁₀OS
:
MFCD02942489
:
118.2 g/mol
:
70°C
Appearance:
White to Almost white powder to crystal
Purity (GC):
min. 98.0 %
Melting point:
48.0 to 52.0 deg-C