
4-Cyanophenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:171364-82-2
:
Formula:C₁₃H₁₆BNO₂
MDL Number:MFCD03093897
:229.09 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:97.0 to 101.0 deg-C