trans,trans-3,4,5-Trifluoro-4'-(4'-propylbicyclohexyl-4-yl)biphenyl ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:137529-41-0
:
Formula:C₂₇H₃₃F₃
MDL Number:MFCD13188632
:414.56 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:101.0 to 105.0 deg-C
- Mesomorphic range:101.0 to 260.0 deg-C
Specification Test Results
Frequently Bought Together