
3-Methylphenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:253342-48-2
:
Formula:C₁₃H₁₉BO₂
MDL Number:MFCD05864303
:218.1 g/mol
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Almost white powder to lump
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:34.0 to 38.0 deg-C