Triethylene Glycol Mono(2-propynyl) ether ≥96.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:208827-90-1
:
Formula:C₉H₁₆O₄
MDL Number:MFCD13189984
:188.22 g/mol
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 96.0 %