Tris Dodecyl sulphate ≥96.0% (by titrimetric analysis) for biochemical research
Supplier: TCI
Certificates
About this item
CAS Number:50996-85-5
:
Formula:C₁₂H₂₆O₄S·C₄H₁₁NO₃
MDL Number:MFCD00043277
:387.53 g/mol
EINECS:256-903-7
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization back titration):min. 96.0 %
- Average molecular weight:384 to 390
- Suitability for protein-analysis:to pass test