Order Entry
Finland
ContactUsLinkComponent
[[4-(Trifluoromethyl)cyclohexyl]methyl]amine(cis- and trans- mixture) ≥97.0% (by GC)
Corrosive
[[4-(Trifluoromethyl)cyclohexyl]methyl]amine(cis- and trans- mixture) ≥97.0% (by GC)
Catalog # TCIAT2896-1G
Supplier:  TCI
CAS Number:  
undefined
Corrosive
[[4-(Trifluoromethyl)cyclohexyl]methyl]amine(cis- and trans- mixture) ≥97.0% (by GC)
Catalog # TCIAT2896-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 97.0 % (total of isomers)
  • Purity (Nonaqueous Titration):
    min. 98.0 %

Specification Test Results

About this item

 :
 :181.20 g/mol
UN:2735
ADR:8,II
 :GHS05
CAS Number:361393-85-3
Formula:C₈H₁₄F₃N
MDL Number:MFCD07777219