Order Entry
Finland
ContactUsLinkComponent
4-(Trifluoromethyl)cyclohexanemethanol (cis- and trans- mixture) cis- and trans-mixture ≥98.0% (by GC)
4-(Trifluoromethyl)cyclohexanemethanol (cis- and trans- mixture) cis- and trans-mixture ≥98.0% (by GC)
Catalog # TCIAT2885-1G
Supplier:  TCI
CAS Number:  
undefined
4-(Trifluoromethyl)cyclohexanemethanol (cis- and trans- mixture) cis- and trans-mixture ≥98.0% (by GC)
Catalog # TCIAT2885-1G
Supplier:  TCI
CAS Number:  

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 % (total of isomers)

Specification Test Results

About this item