Order Entry
Finland
ContactUsLinkComponent
 
 
4-[3-(Trifluoromethyl)-3H-diazirin-3-yl]benzyl bromide ≥95.0% (by HPLC)
 :  TCI
 
undefined
 
4-[3-(Trifluoromethyl)-3H-diazirin-3-yl]benzyl bromide ≥95.0% (by HPLC)
 :  TCI

 

  • Appearance:
    Light yellow to Brown clear liquid
  • Purity (HPLC):
    min. 95.0 area%

 

Frequently Bought Together