L-Tyrosine methyl ester ≥98.0% (by GC, titration analysis)
: TCI
:1080-06-4
:
:C₁₀H₁₃NO₃
:MFCD00002392
:195.22 g/mol
:Refrigerator
:214-095-3
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:133.0 to 137.0 deg-C
- Specific rotation [a]20/D:+26.0 to +29.0 deg (C=1, methanol)