2-Hydroxy-3,5,5-trimethyl-2-cyclohexen-1-one ≥98.0% (by GC)
Catalog # TCIAT2723-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:90.0 to 94.0 deg-C
Specification Test Results
About this item
:
:154.21 g/mol
Boiling Pt:100°C
:GHS07
CAS Number:4883-60-7
Formula:C₉H₁₄O₂
MDL Number:MFCD02181023