
cis-4-(Trifluoromethyl)cyclohexanecarboxylic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:1202578-27-5
:
Formula:C₈H₁₁F₃O₂
MDL Number:MFCD09037361
:196.17 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:97.0 to 101.0 deg-C