



Borane tetrahydrofuran complex 8.5% ∼0.9 mol/L in THF stabilised
Supplier: TCI
Certificates
About this item
CAS Number:14044-65-6
:
Formula:C₄H₁₁BO
MDL Number:MFCD00012429
:85.94 g/mol
Flash Pt:–22°C
Storage Temperature:Refrigerator
EINECS:237-881-8
UN:3399
ADR:4.3,I
:Stabilised with sodium borohydride
:GHS02
:GHS05
:GHS07
:GHS08
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Concentration (Neutralization titration):8.5 to 9.4 w/w%