Order Entry
Finland
ContactUsLinkComponent
Terpin monohydrate ≥98.0% (by GC)
Terpin monohydrate ≥98.0% (by GC)
Catalog # TCIAT2344-25G
Supplier:  TCI
CAS Number:  
undefined
Terpin monohydrate ≥98.0% (by GC)
Catalog # TCIAT2344-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
 :172.27 g/mol
Merck Index:14,09169
CAS Number:2451-01-6
 :MFCD00167312
Formula:C₁₀H₂₀O₂
MDL Number:MFCD00149225