Order Entry
Finland
ContactUsLinkComponent
Tris(2-phenylpyridinato)iridium(III) ≥95.0% (by HPLC), purified by sublimation
Irritant
Tris(2-phenylpyridinato)iridium(III) ≥95.0% (by HPLC), purified by sublimation
Catalog # TCIAT1946-200MG
Supplier:  TCI
CAS Number:  
undefined
Irritant
Tris(2-phenylpyridinato)iridium(III) ≥95.0% (by HPLC), purified by sublimation
Catalog # TCIAT1946-200MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    200 mg
  • Seq:
    2530

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Brown powder to crystal
  • Purity (HPLC):
    min. 95.0 area%

Specification Test Results

About this item

 :
 :654.79 g/mol
 :GHS07
CAS Number:94928-86-6
 :693794-98-8
Formula:C₃₃H₂₄IrN₃
MDL Number:MFCD04039766