3-Bromo-2,2-bis(bromomethyl)propanol ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:1522-92-5
:
Formula:C5H9Br3O
MDL Number:MFCD00021982
:324.8364 g/mol
Boiling Pt:131°C
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Argentmetric Titration):min. 98.0 %
- Melting point:66.0 to 69.0 deg-C
- Solubility in Toluene:almost transparency