Order Entry
Finland
ContactUsLinkComponent
5-(Trimethylsilyl)-1,2,3,4,5-pentamethyl-1,3-cyclopentadiene ≥95.0% (by GC)
Irritant
5-(Trimethylsilyl)-1,2,3,4,5-pentamethyl-1,3-cyclopentadiene ≥95.0% (by GC)
Supplier:  TCI
Certificates
Irritant
5-(Trimethylsilyl)-1,2,3,4,5-pentamethyl-1,3-cyclopentadiene ≥95.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Light yellow to Brown clear liquid
  • Purity (GC):
    min. 95.0 %

Specification Test Results