Order Entry
Finland
ContactUsLinkComponent
Methyl L-tryptophanate hydrochloride ≥98.0% (by HPLC, titration analysis)
Methyl L-tryptophanate hydrochloride ≥98.0% (by HPLC, titration analysis)
Supplier:  TCI
Certificates
Methyl L-tryptophanate hydrochloride ≥98.0% (by HPLC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Argentmetric Titration):
    min. 98.0 %
  • Specific rotation [a]20/D:
    +18.0 to +21.0 deg (C=5, Methanol)

Specification Test Results

Related Information