Order Entry
Finland
ContactUsLinkComponent
 
 
Tris(2,6-dimethoxyphenyl)phosphine ≥97.0% (by HPLC)
 :  TCI
 
undefined
 
Tris(2,6-dimethoxyphenyl)phosphine ≥97.0% (by HPLC)
 :  TCI

 

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (HPLC):
    min. 97.0 area%
  • Melting point:
    153.0 to 157.0 deg-C
  • Lambda max.:
    287.0 to 293.0 nm (MeOH)
  • Absorbance (E1%1cm):
    min. 300 (MeOH, 287.0 to 293.0 nm)