1-(p-Toluenesulphonyl)pyrrole-2-carboxaldehyde ≥98.0% (by titrimetric analysis)
TCIAT1607-5G
: TCI
:
- Pk:5 g
- Seq:3173
- Appearance:White to Yellow to Orange powder to crystal
- Purity (Oxime Formation):min. 98.0 %
- Melting point:94.0 to 97.0 deg-C
:
:249.28 g/mol
:102619-05-6
:C₁₂H₁₁NO₃S
:MFCD00671580