DL-Tryptophan ethyl ester hydrochloride ≥99.0% (by titrimetric analysis)
TCIAT0754-5G
: TCI
:
- Pk:5 g
- Seq:3173
- Appearance:White to Light yellow powder to crystal
- Purity (Argentmetric Titration):min. 99.0 %
- Solubility in Water:almost transparency
:
:268.74 g/mol
:6519-67-1
:C₁₃H₁₆N₂O₂
:MFCD00067553