Order Entry
Finland
ContactUsLinkComponent
7-Hydroxy-4-methylcoumarin-3-acetic acid succinimidyl ester ≥90.0% (by HPLC, total nitrogen)
7-Hydroxy-4-methylcoumarin-3-acetic acid succinimidyl ester ≥90.0% (by HPLC, total nitrogen)
Catalog # TCIAS0960-25MG
Supplier:  TCI
CAS Number:  
undefined
7-Hydroxy-4-methylcoumarin-3-acetic acid succinimidyl ester ≥90.0% (by HPLC, total nitrogen)
Catalog # TCIAS0960-25MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Seq:
    2315

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 90.0 area%
  • Purity (with Total Nitrogen):
    min. 90.0 %
  • Fluorophotometric-detection test:
    to pass test

Specification Test Results

About this item

 :
 :331.28 g/mol
Storage Temperature:Freezer
CAS Number:96735-88-5
Formula:C₁₆H₁₃NO₇
MDL Number:MFCD00467590