

Sulindac Sulphide ≥98.0% (by HPLC)
Catalog # TCIAS0906-25MG
Supplier: TCI
CAS Number:
Specifications
- Pk:25 mg
- Seq:2315
Specifications
Specification Test Results
- Appearance:Light yellow to Brown powder to crystal
- Purity (HPLC):min. 98.0 area%
- Melting point:184.0 to 188.0 deg-C
Specification Test Results
About this item
:
:340.4 g/mol
EINECS:250-892-2
:GHS07
:GHS08
Storage Temperature:Freezer
CAS Number:32004-67-4
Formula:C₂₀H₁₇FO₂S
MDL Number:MFCD00869764