Sulpho-SMCC (3-Sulpho-N-succinimidyl-4-(N-maleimidomethyl)cyclohexane-1-carboxylate sodium salt)
Supplier: TCI
Certificates
About this item
CAS Number:92921-24-9
:
Formula:C₁₆H₁₈N₂O₉S
MDL Number:MFCD00054978
:436.37 g/mol
:GHS07
Storage Temperature:Freezer
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Elemental analysis (Nitrogen):6.20 to 6.70 %