0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Finland
ContactUsLinkComponent
9,9'-Spirobi[9H-fluorene]
9,9'-Spirobi[9H-fluorene] ≥98.0% (by HPLC)
9,9'-Spirobi[9H-fluorene] ≥98.0% (by HPLC)
:
TCI
9,9'-Spirobi[9H-fluorene] ≥98.0% (by HPLC)
:
TCI
:
159-66-0
:
:
C₂₅H₁₆
:
MFCD08704216
:
316.40 g/mol
Appearance:
White to Light yellow powder to crystal
Purity (HPLC):
min. 98.0 area%
Melting point:
200.0 to 204.0 deg-C
Manual Synthetic Organic Chemistry and Materials Chemistry