
Sulphasalazine ≥95.0% (by HPLC, titration analysis)
Catalog # TCIAS0580-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:Light yellow to Amber to Dark green powder to crystal
- Purity (HPLC):min. 95.0 area%
- Purity (Neutralization titration):min. 95.0 %
Specification Test Results
About this item
:
:398.39 g/mol
Merck Index:14,08942
Melting Pt:246°C
EINECS:209-974-3
:GHS08
CAS Number:599-79-1
Formula:C₁₈H₁₄N₄O₅S
MDL Number:MFCD00057363