Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Poly(neopentyl glycol succinate)
Poly(neopentyl glycol succinate)
Poly(neopentyl glycol succinate)
Poly(neopentyl glycol succinate)
Supplier:
TCI
Certificates
Expand image
Poly(neopentyl glycol succinate)
Supplier:
TCI
About this item
CAS Number:
28257-92-3
:
MDL Number:
MFCD00148032
Specification Test Results
Appearance:
White to Light yellow powder to crystal
Suitability for GC-analysis:
to pass test
Specification Test Results
Related Information
Documentation
Manual
Manual Synthetic Organic Chemistry and Materials Chemistry
Documentation