Poly(diethylene glycol succinate)
Catalog # TCIAS0098-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:Colorless to Red to Green clear liquid
- Suitability for GC-analysis:to pass test
Specification Test Results
About this item
:
CAS Number:26183-02-8
MDL Number:MFCD00167429