Ru-Re(FPh)
50 mg
Catalog # TCIAR0100-50MG
Supplier: TCI
Jump to:
Product Details & Documents
- CAS Number:1471276-06-8
- Formula:C₈₆H₇₀F₂₄N₈O₂P₅ReRu
- Molecular Weight:2145.67 g/mol
Documents
Specifications
Specification Test Results
- Appearance:Light yellow to Brown powder to crystal
- Functionality test:to pass test
Specifications
- Pk:50 mg



