
Sodium hydroxymethanesulphinate dihydrate ≥97.0% (by titrimetric analysis)
Catalog # TCIAR0053-500G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
- Seq:3645
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Iodometric Titration):min. 97.0 %
- Solubility in Water:almost transparency
Specification Test Results
About this item
:
:154.12 g/mol
Merck Index:14,08620
EINECS:205-739-4
:GHS07
CAS Number:149-44-0
Formula:CH₃NaO₃S·2H₂O
MDL Number:MFCD00040426