Order Entry
Finland
Orders LinkContactUsLinkComponent
 

MOPS-Na ≥98%

MOPS-Na ≥98%
Certificates
MOPS-Na ≥98%

3 Options Available Below

Product Details & Documents

Purity: 98% CAS No.: 71119-22-7 Molecular Formula: C7H14NNaO4S Formula Weight: 231.25 Physical Form: Cryst powder Appearance: White Melting Point: ca 260° dec. MDL No.: MFCD00064350

Synonyms:  3-(N-Morpholino)propanesulphonic acid sodium salt, Sodium 3-morpholinopropane-1-sulphonate, Sodium 4-morpholin-1-ylpropylsulphonate, MOPS sodium salt
  • CAS Number:71119-22-7
  • Formula:C₇H₁₄NNaO₄S
  • MDL Number:MFCD00064350
  • Molecular Weight:231.25 g/mol
  • Melting Pt:∼ 260 °C (dec.)
  • Storage Temperature:Ambient
  • EINECS:428-420-3
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 77,40 €
    77,40 €
    1 * 25 g
       77,40 €

  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 180,00 €
    180,00 €
    1 * 100 g
       180,00 €

  • Glass bottle for solids
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 676,00 €
    676,00 €
    1 * 500 g
       676,00 €