
Propargyl-PEG3-Carboxylic acid ≥98.0% (by GC, titration analysis)
: TCI
:1347760-82-0
:
:C₁₀H₁₆O₅
:MFCD22201521
:216.23 g/mol
:GHS07
:Freezer
- Appearance:Light yellow to Brown clear liquid
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %