Order Entry
Finland
ContactUsLinkComponent
 
 
1H-Pyrrole-3-carbaldehyde ≥98.0% (by GC)
 :  TCI
 
 
1H-Pyrrole-3-carbaldehyde ≥98.0% (by GC)
 :  TCI

 

  • Appearance:
    White to Brown powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    63.0 to 67.0 deg-C