
3-(2-Pyridyldithio)propionic acid ≥97.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:68617-64-1
:
Formula:C₈H₉NO₂S₂
MDL Number:MFCD18252554
:215.29 g/mol
:GHS07
Storage Temperature:Freezer
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 95.0 area%
- Purity (Neutralization titration):min. 97.0 %
- Melting point:68.0 to 72.0 deg-C
Specification Test Results
Frequently Bought Together