Poly(tert-Butyl Acrylate)-block-Poly(styrene) (Copolymer, 10:25)
Catalog # TCIAP2555-200MG
Supplier: TCI
CAS Number:
Specifications
- Pk:200 mg
- Seq:2530
Specifications
Specification Test Results
- Appearance:White to Yellow to Green powder to crystal
- Molecular weight of number average:33,000 to 38,000
- Molecular weight distribution:1.00 to 1.40
Specification Test Results
About this item
:
CAS Number:697765-49-4