
2-Phenylethylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI
Certificates
About this item
CAS Number:34420-17-2
:
Formula:C₈H₁₁BO₂
MDL Number:MFCD01631226
:149.98 g/mol
Melting Pt:88°C
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):98.0 to 113.0 %
Specification Test Results
Frequently Bought Together