trans,trans-4-n-Propyl-4'-[4-(trifluoromethoxy)phenyl]bicyclohexyl ≥98.0% (by GC)
Catalog # TCIAP2464-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Mesomorphic range:66 to 156 deg-C
Specification Test Results
About this item
:
:368.484 g/mol
CAS Number:133937-72-1
Formula:C₂₂H₃₁F₃O
MDL Number:MFCD20488041