Order Entry
Finland
ContactUsLinkComponent
4-(1-Pyrenyl)phenylboronic acid (contains varying amounts of Anhydride)
4-(1-Pyrenyl)phenylboronic acid (contains varying amounts of Anhydride)
Supplier:  TCI
Certificates
undefined
4-(1-Pyrenyl)phenylboronic acid (contains varying amounts of Anhydride)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Light yellow to Yellow to Green powder to crystal
  • Purity (Neutralization titration):
    95.0 to 105.0 %

Specification Test Results

Related Information

Frequently Bought Together