Order Entry
Finland
ContactUsLinkComponent
Pyromellitic dianhydride ≥99.0% (by HPLC, titration analysis), purified by sublimation
Corrosive
Health
Pyromellitic dianhydride ≥99.0% (by HPLC, titration analysis), purified by sublimation
Supplier:  TCI
Certificates
undefined
Corrosive
Health
Pyromellitic dianhydride ≥99.0% (by HPLC, titration analysis), purified by sublimation
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 99.0 area%
  • Purity (Methanolysis Method):
    min. 99.0 %

Specification Test Results

Related Information

Frequently Bought Together