Order Entry
Finland
ContactUsLinkComponent
Pararosaniline hydrochloride ≥90.0% (by HPLC, total nitrogen) for biochemical research
Health
Pararosaniline hydrochloride ≥90.0% (by HPLC, total nitrogen) for biochemical research
Supplier:  TCI
Certificates
undefined
Health
Pararosaniline hydrochloride ≥90.0% (by HPLC, total nitrogen) for biochemical research
Supplier:  TCI

Specification Test Results

  • Appearance:
    Dark green to Dark blue to Black powder to crystaline
  • Purity (HPLC):
    min. 90.0 area%
  • Purity (with Total Nitrogen):
    min. 90.0 %
  • Lambda max.:
    538.0 to 541.0 nm (EtOH)
  • Absorbance:
    min. 0.400 (2mg/L, EtOH, 538.0 to 541.0 nm) (calcd.on dried substance)

Specification Test Results

Related Information