
4-Propoxyphenylboronic acid (contains varying amounts of Anhydride)
Catalog # TCIAP1968-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):97.0 to 112.0 %
Specification Test Results
About this item
:
:180.01 g/mol
Melting Pt:126°C
:GHS07
CAS Number:186497-67-6
Formula:C₉H₁₃BO₃
MDL Number:MFCD03427189