Pazufloxacin mesylate ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:163680-77-1
:
Formula:C₁₆H₁₅FN₂O₄
MDL Number:MFCD00913262
Melting Pt:259°C
:414.40 g/mol
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Specific rotation [a]20/D:-61.0 to -64.0 deg (C=1, 1mol/L NaOH)