
Phenyl-p-tolylsulphide ≥98.0% (by GC)
Catalog # TCIAP1740-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:Light yellow to Brown clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
About this item
:
:200.30 g/mol
Boiling Pt:312°C
:GHS07
CAS Number:3699-01-2
Formula:C₁₃H₁₂S
MDL Number:MFCD00092378