D-Proline benzyl ester hydrochloride ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI
Certificates
About this item
CAS Number:53843-90-6
:
Formula:C₁₂H₁₅NO₂
MDL Number:MFCD00153458
:241.72 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystaline
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Specific rotation [a]20/D:+42.0 to +46.0 deg (C=1, Methanol)