L-Phenylalanine benzyl ester hydrochloride ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI
Certificates
About this item
CAS Number:2462-32-0
:
Formula:C₁₆H₁₇NO₂
MDL Number:MFCD00043249
:291.78 g/mol
Melting Pt:203°C
EINECS:219-558-3
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Specific rotation [a]20/D:-10.0 to -14.0 deg (C=1, AcOH (80%))