2,5-Dimethyl-1-phenylpyrrole ≥98.0% (by GC)
Catalog # TCIAP1615-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:Light yellow to Brown to Dark green powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:49.0 to 53.0 deg-C
Specification Test Results
About this item
:
:171.24 g/mol
EINECS:201-461-2
CAS Number:83-24-9
Formula:C₁₂H₁₃N
MDL Number:MFCD00022464