0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Finland
ContactUsLinkComponent
2-Phenyl-4-penten-2-ol
2-Phenyl-4-penten-2-ol ≥97.0% (by GC)
2-Phenyl-4-penten-2-ol ≥97.0% (by GC)
TCIAP1330-5G
:
TCI
:
2-Phenyl-4-penten-2-ol ≥97.0% (by GC)
TCIAP1330-5G
:
TCI
:
Pk:
5 g
Seq:
3173
Appearance:
Colorless to Yellow to Orange clear liquid
Purity (GC):
min. 97.0 %
:
:
162.23 g/mol
:
100°C
:
66°C
:
4743-74-2
:
C₁₁H₁₄O
:
MFCD00060886