(S)-2-Hydroxy-3-phenylpropanoic acid ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:20312-36-1
:
Formula:C₉H₁₀O₃
MDL Number:MFCD00004244
:166.17 g/mol
EINECS:243-726-5
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:122.0 to 126.0 deg-C
- Specific rotation [a]20/D:-20.0 to -22.0 deg (C=2, H2O)
Specification Test Results
Frequently Bought Together