Magnesium(II) phthalocyanine
Catalog # TCIAP1018-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Dark red to Dark purple to Dark blue powder to crystal
- Lambda max.:665.0 to 671.0 nm (DMF)
- Molar extinction coefficient:min. 225000 (DMF, 665.0 ~ 671.0 nm)
Specification Test Results
About this item
:
:536.84 g/mol
EINECS:216-766-6
CAS Number:1661-03-6
Formula:C₃₂H₁₆MgN₈
MDL Number:MFCD00041953